missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Hydroxide, Octahydrate, ACS, 98%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor B1040500GM
Specifications
| 12230-71-6 | |
| 500 g | |
| MFCD00149152 | |
| O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++] | |
| 315.46 | |
| ACS |
| 1 | |
| BaH18O10 | |
| ZUDYPQRUOYEARG-UHFFFAOYSA-L | |
| barium(2+) octahydrate dihydroxide | |
| 98% | |
| Poly Bottle |
Chemical Identifiers
| 12230-71-6 | |
| 315.46 | |
| ZUDYPQRUOYEARG-UHFFFAOYSA-L | |
| O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++] |
| BaH18O10 | |
| MFCD00149152 | |
| barium(2+) octahydrate dihydroxide |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.