missing translation for 'onlineSavingsMsg'
Learn More

Barium Hydroxide, Octahydrate, ACS, 98%, Spectrum™ Chemical

Catalog No. p-7165127
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609005 25 g Poly Bottle
18609006 125 g Poly Bottle
18609007 500 g Poly Bottle
18609008 2.5 kg Poly Pail
18609009 12 kg Poly Pail
18609010 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18609005 Supplier Spectrum Chemical Mfg Cor Supplier No. B104025GM
Only null left

Chemical Identifiers

CAS 12230-71-6
Molecular Formula BaH18O10
Molecular Weight (g/mol) 315.46
MDL Number MFCD00149152
InChI Key ZUDYPQRUOYEARG-UHFFFAOYSA-L
IUPAC Name barium(2+) octahydrate dihydroxide
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 98%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.