missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Barium diphenylaminesulfonate, pure, indicator grade

Catalog No. p-3735557
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC401680050 5 g Glass bottle
AC401680100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC401680050 Supplier Thermo Scientific Chemicals Supplier No. 401680050
Only null left

CAS: 6211-24-1 | C24H20BaN2O6S2 | 633.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 6211-24-1
Molecular Formula C24H20BaN2O6S2
Molecular Weight (g/mol) 633.88
MDL Number MFCD00007497
InChI Key IVCNVXFNTKXMCA-UHFFFAOYSA-L
Synonym barium diphenylamine-4-sulfonate, barium diphenylaminesulfonate, benzenesulfonic acid, 4-phenylamino-, barium salt 2:1, barium bis 4-anilinobenzenesulphonate, barium diphenylamine sulfonate, n-phenylsufanilic acid barium, unii-ssj5f79159, diphenylamine-4-sulfonic acid barium salt, n-phenylsufanilic acid, barium salt, 4-anilinobenzenesulfonate; barium 2+
PubChem CID 80342
IUPAC Name 4-anilinobenzenesulfonate;barium(2+)
SMILES [Ba++].[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1.[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Melting Point 300.0°C
Color White to Gray
Loss on Drying 1% max. (105°C)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5 g
Sensitivity (as indicator) passes
Merck Index 15, 7423
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,insoluble in ether and benzene
Formula Weight 633.89
Grade Pure
Physical Form Fine Crystalline Powder
Chemical Name or Material Barium diphenylaminesulfonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON
EINECSNumber 228-278-

RUO – Research Use Only