missing translation for 'onlineSavingsMsg'
Learn More

Barium diphenylamine-4-sulfonate, For Redox Titration, ≥95%, MilliporeSigma™ Supelco™

Catalog No. p-7193491
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
111014921 5 g Poly Bottle
111014919 25 g Poly Bottle
111014920 50 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111014921 Supplier Merck Emd Millipore Supplier No. SGMAD41305G
Only null left

Chemical Identifiers

CAS 6211-24-1
Molecular Formula C24H20BaN2O6S2
Molecular Weight (g/mol) 633.88
MDL Number MFCD00007497
InChI Key IVCNVXFNTKXMCA-UHFFFAOYSA-L
Synonym 4-(Phenylamino)benzenesulfonic acid barium salt; Diphenylamine-4-sulfonic acid barium salt
IUPAC Name barium(2+) bis(4-(phenylamino)benzene-1-sulfonate)
SMILES [Ba++].[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1.[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Quantity 5 g
Linear Formula (C12H10NO3S)2 Ba
Formula Weight 633.88
Percent Purity ≥95%
Packaging Poly Bottle