missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Chloride TS, (U.S.P. Test Solution), Spectrum™ Chemical
B-282, 10326-27-9
$714.27 - $1165.23
Chemical Identifiers
| CAS | 10326-27-9 |
|---|---|
| Molecular Formula | BaCl2H4O2 |
| Molecular Weight (g/mol) | 244.26 |
| MDL Number | MFCD00149154 |
| InChI Key | PWHCIQQGOQTFAE-UHFFFAOYSA-L |
| IUPAC Name | barium(2+) dihydrate dichloride |
| SMILES | O.O.[Cl-].[Cl-].[Ba++] |
Description
Spectrum™ Chemical Barium Chloride TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.Chemical Identifiers
| 10326-27-9 | |
| 244.26 | |
| PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
| O.O.[Cl-].[Cl-].[Ba++] |
| BaCl2H4O2 | |
| MFCD00149154 | |
| barium(2+) dihydrate dichloride |
Specifications
| 10326-27-9,99.9% | |
| 100 mL | |
| MFCD00149154 | |
| O.O.[Cl-].[Cl-].[Ba++] | |
| 244.26 |
| 0.1%,99.9% | |
| BaCl2H4O2 | |
| PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
| barium(2+) dihydrate dichloride | |
| Poly Bottle |