missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride TS, (U.S.P. Test Solution), Spectrum™ Chemical

Catalog No. 18600845
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600845 100 mL Poly Bottle
18600846 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18600845 Supplier Spectrum Chemical Mfg Cor Supplier No. B282100ML
Only null left

B-282, 10326-27-9

Spectrum™ Chemical Barium Chloride TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

Chemical Identifiers

CAS 10326-27-9 , 99.9%
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
IUPAC Name barium(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

CAS Max % 0.1%, 99.9%
Quantity 100 mL
Packaging Poly Bottle