missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium chloride dihydrate, Puriss. p.a. plus, ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Honeywell Fluka™
Puriss. p.a. plus, ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99%
Supplier: Honeywell-Fluka 311254X5KG
Description
- Exceeds standard ACS specifications with additional trace metals analysis
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
| Barium Chloride Dihydrate | |
| 10326-27-9 | |
| BaCl2H4O2 | |
| UN1564 | |
| PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
| barium(2+);dichloride;dihydrate | |
| 5284346 | |
| 244.26 | |
| ACS/puriss. p.a. plus |
| Reag. Ph. Eur. reag. ISO | |
| 4 x 5 kg | |
| MFCD00149154 | |
| barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
| O.O.[Cl-].[Cl-].[Ba++] | |
| 244.26 | |
| CHEBI:86153 | |
| ≥99% |
Chemical Identifiers
| 10326-27-9 | |
| 244.26 | |
| PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
| 5284346 | |
| barium(2+);dichloride;dihydrate |
| BaCl2H4O2 | |
| MFCD00149154 | |
| barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
| CHEBI:86153 | |
| O.O.[Cl-].[Cl-].[Ba++] |
Safety and Handling
Missing Phrase - N15.00950417-P261
H301-H332
DOTInformation : 6.1 / PGIII
EINECSNumber : 233-788-1
RTECSNumber : CQ8751000