missing translation for 'onlineSavingsMsg'
Learn More

Barium chloride dihydrate, Puriss. p.a. plus, ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Honeywell Fluka™

Catalog No. p-7071996
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
4 x 5 kg
6 x 1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000360 100 g
6000363 500 g
6000361 1 kg
60045910 6 x 1 kg
60045909 4 x 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 6000360 Supplier Honeywell Chemicals Supplier No. 31125100G
Only null left

Puriss. p.a. plus, ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99%

  • Exceeds standard ACS specifications with additional trace metals analysis
  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Chemical Identifiers

CAS 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PubChem CID 5284346
ChEBI CHEBI:86153
IUPAC Name barium(2+);dichloride;dihydrate
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

Quantity 100 g
UN Number 1564
Formula Weight 244.26g/mol
Percent Purity ≥99%
Grade ACS/puriss. p.a. plus
Packaging Plastic Bottle
Chemical Name or Material Barium Chloride Dihydrate, Reag. Ph. Eur. reag. ISO
Hazard Statement P261
Precautionary Statement H301-H332
EINECSNumber 233-788-1
RTECSNumber CQ8751000