missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Chloride, Dihydrate, Crystal, ACS, 99%, Spectrum™ Chemical
$308.69 - $6020.38
Chemical Identifiers
| CAS | 10326-27-9 |
|---|---|
| Molecular Formula | BaCl2H4O2 |
| Molecular Weight (g/mol) | 244.26 |
| MDL Number | MFCD00149154 |
| InChI Key | PWHCIQQGOQTFAE-UHFFFAOYSA-L |
| IUPAC Name | barium(2+) dihydrate dichloride |
| SMILES | O.O.[Cl-].[Cl-].[Ba++] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18608985
|
Spectrum Chemical Mfg Cor
B1025125GM |
125 g | Poly Bottle |
Each for $308.69
|
|
||||
|
18608986
|
Spectrum Chemical Mfg Cor
B1025500GM |
500 g | Poly Bottle |
Each for $477.72
|
|
||||
|
18608987
|
Spectrum Chemical Mfg Cor
B102525KG |
2.5 kg | Poly Bottle |
Each for $1,423.22
|
|
||||
|
18608988
|
Spectrum Chemical Mfg Cor
B102512KG |
12 kg | Poly Pail |
Each for $2,752.28
|
|
||||
|
18608989
|
Spectrum Chemical Mfg Cor
B102550KGBL |
50 kg | Fiber Drum |
Each for $6,020.38
|
|
||||
Chemical Identifiers
| 10326-27-9 | |
| 244.26 | |
| PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
| O.O.[Cl-].[Cl-].[Ba++] |
| BaCl2H4O2 | |
| MFCD00149154 | |
| barium(2+) dihydrate dichloride |
Specifications
| 10326-27-9 | |
| 5.2 to 8.2 | |
| BaCl2H4O2 | |
| PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
| barium(2+) dihydrate dichloride | |
| 99% | |
| 14.0-16.0% |
| 1 | |
| 125 g | |
| MFCD00149154 | |
| O.O.[Cl-].[Cl-].[Ba++] | |
| 244.26 | |
| ACS | |
| Poly Bottle |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.