missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Chloride, Dihydrate, Crystal, ACS, 99%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor B102512KG
Specifications
| 10326-27-9 | |
| 5.2 to 8.2 | |
| BaCl2H4O2 | |
| PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
| barium(2+) dihydrate dichloride | |
| 99% | |
| 14.0-16.0% |
| 1 | |
| 12 kg | |
| MFCD00149154 | |
| O.O.[Cl-].[Cl-].[Ba++] | |
| 244.26 | |
| ACS | |
| Poly Pail |
Chemical Identifiers
| 10326-27-9 | |
| 244.26 | |
| PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
| O.O.[Cl-].[Cl-].[Ba++] |
| BaCl2H4O2 | |
| MFCD00149154 | |
| barium(2+) dihydrate dichloride |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.