missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride, Certified, 0.100N ±0.001N (0.05M), LabChem™

Catalog No. LC116204
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC116204 4 L
LC116202 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC116204 Supplier LabChem Supplier No. LC116204
Only null left

Chemical Identifiers

CAS 10326-27-9 , 7732-18-5
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PubChem CID 5284346
ChEBI CHEBI:86153
IUPAC Name barium(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

Color Colorless
CAS Max % 98.78, 1.22
Physical Form Liquid
Quantity 4 L
Linear Formula BaCl2·2H2O
Solubility Information Soluble in water
Formula Weight 244.26
Grade Certified
Concentration or Composition (by Analyte or Components) 0.100N ±0.001N (0.05M)
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Hydrogen chloride; Barium
Chemical Name or Material Barium Chloride
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature