missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride, 5% w/v in 0.5 N Hydrochloric Acid, Reagents

Catalog No. 01335854
Change view
Click to view available options
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01335854 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335854 Supplier Labchem Inc Supplier No. CB0025004A

Barium Chloride, 5% w/v in 0.5 N Hydrochloric Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 10326-27-9 , 10326-27-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
pH <1
Physical Form Liquid
Quantity 4 L
Percent Purity 5%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.0447
Chemical Name or Material Barium Chloride, 5% w/v in 0.5 N Hydrochloric Acid