missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride, 25% (w/v), Ricca Chemical

Catalog No. 86016
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
Catalog No. Quantity
86016 500 mL
1 options

Catalog No. 86016

Supplier: Ricca Chemical Company 86016

Only null left

Chemical Identifiers

CAS 7732-18-5 , 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PubChem CID 5284346
ChEBI CHEBI:86153
IUPAC Name barium(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

Physical Form Liquid
Quantity 500 mL
Percent Purity 25% W/V
Grade Laboratory
Packaging Natural Poly Bottle
Chemical Name or Material Barium chloride