missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride, 2.8% w/v in 5.6% v/v Hydrochloric Acid, Reagents

Catalog No. 01335799
Change view
Click to view available options
Quantity: 20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01335799 Quantity: 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335799 Supplier Labchem Inc Supplier No. CB11050020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Barium Chloride, 2.8% w/v in 5.6% v/v Hydrochloric Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 10326-27-9 , 10326-27-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
pH <1
Physical Form Liquid
Quantity 20 L
Percent Purity 2.85%
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.05
Chemical Name or Material Barium Chloride, 2.8% w/v in 5.6% v/v Hydrochloric Acid