missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Chloride, 11.8% (w/v) (118 g/L), Ricca Chemical
Supplier: Ricca Chemical Company 85216
Specifications
| Barium Chloride | |
| 88.0,11.0 | |
| BaCl2H4O2 | |
| PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
| barium(2+);dichloride;dihydrate | |
| 5284346 | |
| Natural Poly Bottle | |
| 500 mL |
| 10326-27-9 , 7732-18-5 | |
| 91.0,12.0 | |
| barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
| O.O.[Cl-].[Cl-].[Ba+2] | |
| 244.257 | |
| CHEBI:86153 | |
| Aqueous Solution | |
| 11.8% (w/v); 118g/L |
Chemical Identifiers
| 10326-27-9 | |
| 244.257 | |
| barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
| CHEBI:86153 | |
| O.O.[Cl-].[Cl-].[Ba+2] |
| BaCl2H4O2 | |
| PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
| 5284346 | |
| barium(2+);dichloride;dihydrate |