missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride, 10% (w/v), Ricca Chemical

Supplier:  Ricca Chemical Company R08500004F

 View more versions of this product

Catalog No. R08500004F


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Barium Chloride
10326-27-9,7732-18-5
93.0,11.0
MFCD00149154
PWHCIQQGOQTFAE-UHFFFAOYSA-L
barium(2+) dihydrate dichloride
5284346
10%
Cubitainer
6.5 to 6.9
Liquid
Colorless
89.0,9.0
BaCl2H4O2
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
O.O.[Cl-].[Cl-].[Ba++]
244.26
CHEBI:86153
Laboratory
Aqueous Solution
4 L
10% (w/v)

Chemical Identifiers

10326-27-9
244.26
PWHCIQQGOQTFAE-UHFFFAOYSA-L
5284346
barium(2+) dihydrate dichloride
BaCl2H4O2
MFCD00149154
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
CHEBI:86153
O.O.[Cl-].[Cl-].[Ba++]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions