missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Chloride, 10% (w/v), Ricca Chemical
Supplier: Ricca Chemical Company 85032
Specifications
| Barium Chloride | |
| 10326-27-9,7732-18-5 | |
| 93.0,11.0 | |
| MFCD00149154 | |
| PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
| barium(2+) dihydrate dichloride | |
| 5284346 | |
| 10% | |
| Natural Poly Bottle | |
| 6.5 to 6.9 | |
| Liquid |
| Colorless | |
| 89.0,9.0 | |
| BaCl2H4O2 | |
| barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
| O.O.[Cl-].[Cl-].[Ba++] | |
| 244.26 | |
| CHEBI:86153 | |
| Laboratory | |
| Aqueous Solution | |
| 1 L | |
| 10% (w/v) |
Chemical Identifiers
| 10326-27-9 | |
| 244.26 | |
| PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
| 5284346 | |
| barium(2+) dihydrate dichloride |
| BaCl2H4O2 | |
| MFCD00149154 | |
| barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
| CHEBI:86153 | |
| O.O.[Cl-].[Cl-].[Ba++] |