missing translation for 'onlineSavingsMsg'
Learn More

Barium chloride, 0.1N Standardized Solution

Catalog No. p-7050095
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35582K2 1 L
AA35582K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA35582K2 Supplier Thermo Scientific Chemicals Supplier No. 035582K2
Only null left

CAS: 10361-37-2 | BaCl2 | 208.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10361-37-2
Molecular Formula BaCl2
Molecular Weight (g/mol) 208.23
MDL Number MFCD00003445
InChI Key WDIHJSXYQDMJHN-UHFFFAOYSA-L
Synonym barium chloride, barium dichloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508
PubChem CID 25204
ChEBI CHEBI:63317
IUPAC Name barium(2+);dichloride
SMILES [Cl-].[Cl-].[Ba++]

Specifications

Color Colorless
Concentration 0.1N
Formula Weight 208.25
Odor Odorless
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Barium chloride, Standardized Solution
EINECSNumber 233-788-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only