missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloranilate Trihydrate 97.0+%, TCI America™

Catalog No. c007825g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C007825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C007825G Supplier TCI America Supplier No. C007825G
Only null left

Chemical Identifiers

CAS 13435-46-6
Molecular Formula C6BaCl2O4
Molecular Weight (g/mol) 344.289
MDL Number MFCD00078327
InChI Key IPBSAQBBXCGJQG-UHFFFAOYSA-L
Synonym Chloranilic Acid Barium Salt, Barium 3,6-Dichloro-1,4-dioxo-3,6-cyclohexadiene-2,5-diolate
PubChem CID 83446
IUPAC Name barium(2+);2,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,4-diolate
SMILES C1(=C(C(=O)C(=C(C1=O)Cl)[O-])Cl)[O-].[Ba+2]

Specifications

Color Black-Purple
Quantity 25 g
UN Number 1564
Formula Weight 344.29
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Barium Chloranilate Trihydrate