CAS RN 13435-46-6
CAS RN 13435-46-6
IUPAC Name:
barium(2+) 2,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,4-bis(olate)
Synonyms:
barium(2+) 2,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,4-bis(olate)
Molecular Weight (g/mol):
344.29
Molecular Formula:
C6BaCl2O4
InChi Key:
IPBSAQBBXCGJQG-UHFFFAOYSA-L
SMILES:
[Ba++].[O-]C1=C(Cl)C(=O)C([O-])=C(Cl)C1=O
1
–
1
of
1
results
1
Barium Chloranilate Trihydrate 97.0+%, TCI America™
CAS: 13435-46-6 Molecular Formula: C6BaCl2O4 Molecular Weight (g/mol): 344.289 MDL Number: MFCD00078327 InChI Key: IPBSAQBBXCGJQG-UHFFFAOYSA-L Synonym: Chloranilic Acid Barium Salt, Barium 3,6-Dichloro-1,4-dioxo-3,6-cyclohexadiene-2,5-diolate PubChem CID: 83446 IUPAC Name: barium(2+);2,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,4-diolate SMILES: C1(=C(C(=O)C(=C(C1=O)Cl)[O-])Cl)[O-].[Ba+2]