missing translation for 'onlineSavingsMsg'
Learn More

Barium carbonate, tech.

Used in the manufacture of glass, brick, clay products, barium chemicals, barium ferrites, and production of photographic papers | CAS: 513-77-9 | CBaO3 | 197.34 g/mol

Supplier:  Thermo Scientific Chemicals 043478A9

 View more versions of this product

Catalog No. AA43478A9


Only null left
Explore available promotions

Description

Description

Used in the manufacture of glass, brick, clay products, barium chemicals, barium ferrites, and production of photographic papers.Barium carbonate has been used as brine purification chemical in the chloro-alkali industry and as an ingredient in rat poisons and enamels. Barium carbonate is used in photographic paper manufacturing. The chemical is used to produce the barium sulphate which gives the photographic paper its flat white appearance. Practiced in the manufacture of barium ferrites and also in the manufacture of television picture tubes. Barium carbonate is an additive in oil-well drilling muds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(decomposition)
811°C
White
10 kg
MFCD00003448
Almost insoluble in water. Soluble in NH4Cl solution,dilute HCl,HNO3,acetic acid. Insoluble in most alkalies.Soluble in most acids,with the exception of sulfuric acid. It is insoluble in water and ethanol.
[Ba++].[O-]C([O-])=O
197.34
197.35
4.43 g/cm3
Barium carbonate
513-77-9
Powder
CBaO3
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
AYJRCSIUFZENHW-UHFFFAOYSA-L
barium(2+);carbonate
10563
Odorless

Chemical Identifiers

513-77-9
197.34
AYJRCSIUFZENHW-UHFFFAOYSA-L
10563
CBaO3
MFCD00003448
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
[Ba++].[O-]C([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Avoid contact with skin and eyes.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 208-167-3

RTECSNumber : CQ8600000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only