missing translation for 'onlineSavingsMsg'
Learn More

Barium Carbonate, Honeywell Fluka™

Puriss. p.a., ACS reagent

$554.79 - $554.79

Chemical Identifiers

CAS 513-77-9
Molecular Formula CBaO3
Molecular Weight (g/mol) 197.34
MDL Number MFCD00003448
InChI Key AYJRCSIUFZENHW-UHFFFAOYSA-L
Synonym barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
PubChem CID 10563
SMILES [Ba++].[O-]C([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6001911
View Documents
Honeywell Chemicals
117291KG
1 kg
Each for $554.79
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

513-77-9
197.34
AYJRCSIUFZENHW-UHFFFAOYSA-L
10563
CBaO3
MFCD00003448
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
[Ba++].[O-]C([O-])=O

Specifications

513-77-9
CBaO3
MFCD00003448
7045119
AYJRCSIUFZENHW-UHFFFAOYSA-L
barium(2+);carbonate
10563
ACS/puriss. p.a.
1 kg
BaCO3
NONH for all modes of transport
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
[Ba++].[O-]C([O-])=O
197.34
197.34g/mol
Barium Carbonate
Videos
Safety and Handling

Safety and Handling

P301 + P312 + P330

H302

EINECSNumber : 208-167-3

RTECSNumber : CQ8600000

SDS
Documents

Documents