missing translation for 'onlineSavingsMsg'
Learn More

Barium carbonate, ACS, 99.0-101.0%

Catalog No. p-4407257
Change view
Click to view available options
Quantity:
500 g
1 kg
2 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3623636 500 g
AA36236A1 1 kg
AA36236D3 2 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3623636 Supplier Thermo Scientific Chemicals Supplier No. 03623636
Only null left

CAS: 513-77-9 | CBaO3 | 197.34 g/mol

Barium carbonate has been used as brine purification chemical in the chloro-alkali industry and as an ingredient in rat poisons and enamels. Barium carbonate is used in photographic paper manufacturing. The chemical is used to produce the barium sulphate which gives the photographic paper its flat white appearance. Practiced in the manufacture of barium ferrites and also in the manufacture of television picture tubes. Barium carbonate is an additive in oil-well drilling muds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 513-77-9
Molecular Formula CBaO3
Molecular Weight (g/mol) 197.34
MDL Number MFCD00003448
InChI Key AYJRCSIUFZENHW-UHFFFAOYSA-L
Synonym barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
PubChem CID 10563
SMILES [Ba++].[O-]C([O-])=O

Specifications

Boiling Point (decomposition)
Melting Point 811°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.0 to 101.0%
Solubility Information Soluble in most acids,with the exception of sulfuric acid. It is insoluble in water and ethanol.
IUPAC Name barium(2+) carbonate
Formula Weight 197.35
Grade ACS
Odor Odorless
Appearance White powder
Density 4.43 g/cm3
Chemical Name or Material Barium carbonate, For analysis ACS
Show More Show Less
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 208-167-3
RTECSNumber CQ8600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only