missing translation for 'onlineSavingsMsg'
Learn More

Barium Carbonate, ACS, 99.0-101.0%, Spectrum™ Chemical

Catalog No. p-7165120
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608968 125 g Poly Bottle
18608970 500 g Poly Bottle
18608972 2.5 kg Poly Bottle
18608971 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18608968 Supplier Spectrum Chemical Mfg Cor Supplier No. B1015125GM
Only null left

Chemical Identifiers

CAS 513-77-9
Molecular Formula CBaO3
Molecular Weight (g/mol) 197.34
MDL Number MFCD00003448
InChI Key AYJRCSIUFZENHW-UHFFFAOYSA-L
IUPAC Name barium(2+) carbonate
SMILES [Ba++].[O-]C([O-])=O

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 99 to 101%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.