Learn More
Barium carbonate, 99%
Description
Barium carbonate is widely used in the ceramics industry as an ingredient in glazes. It acts as a flux, a matting and crystallizing agent and combines with certain colouring oxides to produce unique colours not easily attainable by other means. It is also used for removing of sulfates in phosphoric acid production and chlorine alkali electrolysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Barium carbonate is widely used in the ceramics industry as an ingredient in glazes. It acts as a flux, a matting and crystallizing agent and combines with certain colouring oxides to produce unique colours not easily attainable by other means. It is also used for removing of sulfates in phosphoric acid production and chlorine alkali electrolysis.
Solubility
Insoluble in water and alcohol. Soluble in acetic acid, hydrochloric acid and nitric acid.
Notes
m.p. 174° (90 atm), 811° (1 atm)
Specifications
Specifications
| Boiling Point | (decomposition) |
| Melting Point | 811°C |
| Quantity | 25 g |
| Linear Formula | BaCO3 |
| Merck Index | 14,969 |
| Solubility Information | Insoluble in water and alcohol. Soluble in acetic acid,hydrochloric acid and nitric acid. |
| IUPAC Name | barium(2+) carbonate |
| Formula Weight | 197.35 |
| Percent Purity | 99% |
| Odor | Odorless |
| Show More |
Safety and Handling
| Hazard Category | H302 |
| Hazard Statement | GHS H Statement H302 Harmful if swallowed. |
| Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
| EINECSNumber | 208-167-3 |
| RTECSNumber | CQ8600000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only