Learn More
Barium carbonate, 99.95% (metals basis)
CAS: 513-77-9 | CBaO3 | 197.34 g/mol
Supplier: Thermo Scientific Chemicals 01445218
Description
Barium carbonate is frequently employed in ceramic industry as a component of glazes. It is added to clay to precipitate some soluble salts while manufacturing bricks, tiles and potteries. Barium carbonate is mainly used as a raw material in the electrical engineering industries for glass production, and as a raw material in the magnet manufacturing industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (decomposition) | |
| 811°C | |
| White | |
| 50 g | |
| CBaO3 | |
| barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p | |
| AYJRCSIUFZENHW-UHFFFAOYSA-L | |
| barium(2+) carbonate | |
| 10563 | |
| Odorless | |
| 4.43 |
| Barium carbonate | |
| 513-77-9 | |
| Lump/Powder | |
| 99.95% | |
| MFCD00003448 | |
| Insoluble in water. Soluble in most of the acids,with the exception of sulfuric acid. | |
| [Ba++].[O-]C([O-])=O | |
| 197.34 | |
| 197.35 | |
| Off-white lump/powder |
Chemical Identifiers
| 513-77-9 | |
| 197.34 | |
| AYJRCSIUFZENHW-UHFFFAOYSA-L | |
| 10563 |
| CBaO3 | |
| MFCD00003448 | |
| barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p | |
| [Ba++].[O-]C([O-])=O |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 208-167-3
RTECSNumber : CQ800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only