missing translation for 'onlineSavingsMsg'
Learn More

Barium bromide dihydrate, 99.3%

CAS: 7791-28-8 | BaBr2H4O2 | 333.17 g/mol

$116.51 - $289.51

Chemical Identifiers

CAS 7791-28-8
Molecular Formula BaBr2H4O2
Molecular Weight (g/mol) 333.17
MDL Number MFCD00149153
InChI Key HQQMRSGBIPFNSN-UHFFFAOYSA-L
IUPAC Name barium(2+) dihydrate dibromide
SMILES O.O.[Br-].[Br-].[Ba++]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1219722
View Documents
Thermo Scientific Chemicals
01219722
100 g
Each for $116.51
Only null left
 
AA1219736
View Documents
Thermo Scientific Chemicals
01219736
500 g
Each for $289.51
Only null left
 
Description

Description

Barium bromide dihydrate is used to purify radium and make zinc bromide. It also used  precursor to chemicals used in photograph.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7791-28-8
333.17
HQQMRSGBIPFNSN-UHFFFAOYSA-L
O.O.[Br-].[Br-].[Ba++]
BaBr2H4O2
MFCD00149153
barium(2+) dihydrate dibromide

Specifications

7791-28-8
100 g
BaBr2H4O2
Very soluble in water,soluble in methanol
O.O.[Br-].[Br-].[Ba++]
333.17
3.58
Crystalline
99.30%
MFCD00149153
HQQMRSGBIPFNSN-UHFFFAOYSA-L
barium(2+) dihydrate dibromide
333.17 (297.14 Anhydrous)
Barium bromide dihydrate
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 234-140-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only