Learn More
Barium bromide dihydrate, 98+%
CAS: 7791-28-8 | BaBr2H4O2 | 333.17 g/mol
$183.59 - $183.59
Chemical Identifiers
| CAS | 7791-28-8 |
|---|---|
| Molecular Formula | BaBr2H4O2 |
| Molecular Weight (g/mol) | 333.17 |
| MDL Number | MFCD00149153 |
| InChI Key | HQQMRSGBIPFNSN-UHFFFAOYSA-L |
| IUPAC Name | barium(2+) dihydrate dibromide |
| SMILES | O.O.[Br-].[Br-].[Ba++] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1632836
|
Thermo Scientific Chemicals
A1632836 |
500 g |
Each for $183.59
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7791-28-8 | |
| 333.17 | |
| HQQMRSGBIPFNSN-UHFFFAOYSA-L | |
| O.O.[Br-].[Br-].[Ba++] |
| BaBr2H4O2 | |
| MFCD00149153 | |
| barium(2+) dihydrate dibromide |
Specifications
| 7791-28-8 | |
| BaBr2H4O2 | |
| UN1564 | |
| HQQMRSGBIPFNSN-UHFFFAOYSA-L | |
| barium(2+) dihydrate dibromide | |
| 333.17 (297.15 Anhydrous) | |
| Hygroscopic | |
| Barium bromide dihydrate |
| 500 g | |
| MFCD00149153 | |
| 14,968 | |
| O.O.[Br-].[Br-].[Ba++] | |
| 333.17 | |
| ≥98% | |
| 3.58 |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
EINECSNumber : 234-140-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only