Learn More
Barium acetate, 99+%, for analysis
CAS: 543-80-6 | C4H6BaO4 | 255.42 g/mol
Supplier: Thermo Scientific Chemicals 222800010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Barium acetate | |
| 543-80-6 | |
| 7 to 8.5 (at 5% 20°C) | |
| 1 kg | |
| C4H6BaO4 | |
| MFCD00012447 | |
| barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate | |
| ITHZDDVSAWDQPZ-UHFFFAOYSA-L | |
| barium(2+) diacetate | |
| 10980 | |
| ≥99% | |
| Plastic bottle | |
| 2.4600g/mL |
| For analysis ACS | |
| White | |
| Crystalline Powder | |
| 99.0 to 102% (Complexometry) | |
| (CH3CO2)2Ba | |
| 15, 961 | |
| Solubility in water: soluble. Other solubilities: practically insoluble in alcohol | |
| [Ba++].CC([O-])=O.CC([O-])=O | |
| 255.42 | |
| 255.43 | |
| Analytical | |
| 2.46 |
Chemical Identifiers
| 543-80-6 | |
| 255.42 | |
| ITHZDDVSAWDQPZ-UHFFFAOYSA-L | |
| 10980 | |
| [Ba++].CC([O-])=O.CC([O-])=O |
| C4H6BaO4 | |
| MFCD00012447 | |
| barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate | |
| barium(2+) diacetate |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 208-849-
RUO – Research Use Only