missing translation for 'onlineSavingsMsg'
Learn More

Barium acetate, 99+%, for analysis

Catalog No. p-3776849
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC222805000 500 g Plastic bottle
AC222800010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC222805000 Supplier Thermo Scientific Chemicals Supplier No. 222805000
Only null left

CAS: 543-80-6 | C4H6BaO4 | 255.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 543-80-6
Molecular Formula C4H6BaO4
Molecular Weight (g/mol) 255.42
MDL Number MFCD00012447
InChI Key ITHZDDVSAWDQPZ-UHFFFAOYSA-L
Synonym barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate
PubChem CID 10980
IUPAC Name barium(2+) diacetate
SMILES [Ba++].CC([O-])=O.CC([O-])=O

Specifications

Color White
pH 7 to 8.5 (at 5% 20°C)
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99.0 to 102% (Complexometry)
Linear Formula (CH3CO2)2Ba
Merck Index 15, 961
Solubility Information Solubility in water: soluble. Other solubilities: practically insoluble in alcohol
Formula Weight 255.43
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 2.46
Density 2.4600g/mL
Chemical Name or Material Barium acetate, For analysis ACS
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-849-

RUO – Research Use Only