missing translation for 'onlineSavingsMsg'
Learn More

Barium 2-ethylhexanoate, 20% w/w in 2-ethylhexanoic acid, Thermo Scientific™

$78.18 - $1055.01

Chemical Identifiers

CAS 2457-01-4
MDL Number MFCD00058696
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4402109
View Documents
Thermo Scientific Chemicals
4402109
10 g
Each for $78.18
Only null left
 
AA4402118
View Documents
Thermo Scientific Chemicals
4402118
50 g
Each for $276.81
Only null left
 
AA4402130
View Documents
Thermo Scientific Chemicals
4402130
250 g
Each for $1,055.01
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2457-01-4
MFCD00058696

Specifications

2457-01-4
C16H30O4Sr
10 g
DLWBHRIWCMOQKI-UHFFFAOYNA-L
strontium;2-ethylhexanoate
102822
20% w/w in 2-ethylhexanoic acid
Barium 2-ethylhexanoate
Weak, characteristic
MFCD00058696
strontium 2-ethylhexanoate, strontium bis 2-ethylhexanoate, hexanoic acid, 2-ethyl-, strontium salt 2:1, acmc-20ajmv, bis 2-ethylhexanoic acid strontium salt, hexanoic acid, 2-ethyl-, strontium salt, strontium 2+ bis 2-ethylhexanoate, hexanoic acid,2-ethyl-, strontium salt 2:1
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Sr+2]
374.03
423.76
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361-H302-H332
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful if inhaled.

P201-P202-P281-P308+P313-P501c

H361d

EINECSNumber : 219-535-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents