accessibility menu, dialog, popup

UserName

CAS RN 2457-01-4

IUPAC Name: barium(2+) bis(2-ethylhexanoate)
Synonyms: barium(2+) bis(2-ethylhexanoate)
Molecular Weight (g/mol): 423.74
Molecular Formula: C16H30BaO4
InChi Key: VJFFDDQGMMQGTQ-UHFFFAOYNA-L
SMILES: [Ba++].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)