missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium 2-Cyanoethylphosphate Hydrate 98.0+%, TCI America™
$123.03 - $359.76
Chemical Identifiers
| CAS | 5015-38-3 |
|---|---|
| Molecular Formula | C3H4BaNO4P |
| Molecular Weight (g/mol) | 286.369 |
| MDL Number | MFCD00012588 |
| InChI Key | MRQIDZJGQMWVQR-UHFFFAOYSA-L |
| Synonym | 2-Cyanoethylphosporic Acid Barium Salt |
| PubChem CID | 78714 |
| IUPAC Name | barium(2+);2-cyanoethyl phosphate |
| SMILES | C(COP(=O)([O-])[O-])C#N.[Ba+2] |
Chemical Identifiers
| 5015-38-3 | |
| 286.369 | |
| MRQIDZJGQMWVQR-UHFFFAOYSA-L | |
| 78714 | |
| C(COP(=O)([O-])[O-])C#N.[Ba+2] |
| C3H4BaNO4P | |
| MFCD00012588 | |
| 2-Cyanoethylphosporic Acid Barium Salt | |
| barium(2+);2-cyanoethyl phosphate |
Specifications
| 5015-38-3 | |
| C3H4BaNO4P | |
| 5 g | |
| 2-Cyanoethylphosporic Acid Barium Salt | |
| C(COP(=O)([O-])[O-])C#N.[Ba+2] | |
| 286.369 | |
| 286.37 | |
| Crystalline Powder |
| White | |
| MFCD00012588 | |
| 1564 | |
| MRQIDZJGQMWVQR-UHFFFAOYSA-L | |
| barium(2+);2-cyanoethyl phosphate | |
| 78714 | |
| ≥98.0% (T) | |
| Barium 2-Cyanoethylphosphate Hydrate [Phosphorylating Agent] |
Safety and Handling
TSCA : Yes