missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium 2-Cyanoethylphosphate Hydrate 98.0+%, TCI America™
Supplier: TCI America C09785G
Specifications
| Barium 2-Cyanoethylphosphate Hydrate [Phosphorylating Agent] | |
| White | |
| MFCD00012588 | |
| 1564 | |
| MRQIDZJGQMWVQR-UHFFFAOYSA-L | |
| barium(2+);2-cyanoethyl phosphate | |
| 78714 | |
| ≥98.0% (T) |
| 5015-38-3 | |
| C3H4BaNO4P | |
| 5 g | |
| 2-Cyanoethylphosporic Acid Barium Salt | |
| C(COP(=O)([O-])[O-])C#N.[Ba+2] | |
| 286.369 | |
| 286.37 | |
| Crystalline Powder |
Chemical Identifiers
| 5015-38-3 | |
| 286.369 | |
| MRQIDZJGQMWVQR-UHFFFAOYSA-L | |
| 78714 | |
| C(COP(=O)([O-])[O-])C#N.[Ba+2] |
| C3H4BaNO4P | |
| MFCD00012588 | |
| 2-Cyanoethylphosporic Acid Barium Salt | |
| barium(2+);2-cyanoethyl phosphate |
Safety and Handling
TSCA : Yes