Learn More
Aspen Surgical™ Bard-Parker™ Surgical Blades
Offer consistent sharpness, control and strength
$191.00 - $553.57
Specifications
| Blade Material | Carbon Steel |
|---|---|
| Product Type | Surgical Blade |
| For Use With (Application) | For Tissue Separation, and Other Procedures that require a Sharp Surgical Blade to Puncture or Cut |
| Catalog Number | Mfr. No. | Size | Sterility | Handle Type | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Sterility | Handle Type | Price | Quantity & Availability | |||
|
0891511
|
Aspen Surgical™
371121 |
# 21 | Sterile | Ribbed |
Case of 150 Each for $553.57
|
|
|||
|
08914A
|
Aspen Surgical™
371310 |
# 10 | Non-sterile | Ribbed |
Case of 150 Each for $501.04
|
|
|||
|
08914B
|
Aspen Surgical™
371311 |
# 11 | Non-sterile | Ribbed |
Case of 150 Each for $501.04
|
|
|||
|
089151B
|
Aspen Surgical™
371322 |
# 22 | Non-sterile | Ribbed |
Case of 150 Each for $528.71
|
|
|||
|
089151C
|
Aspen Surgical™
371325 |
# 25 | Non-sterile | Ribbed |
Case of 150 Each for $528.71
|
|
|||
|
089159
|
Aspen Surgical™
371115 |
# 15 | Sterile | Ribbed |
Case of 150 Each for $553.57
|
|
|||
|
22027982
|
Aspen Surgical™
371122 |
# 22 | Sterile | Ribbed |
Pack of 50 Each for $191.00
Case of 3 Pack for $573.00
|
|
|||
Description
Offer consistent sharpness, control and strength
- Each blade packaged in foil pouch with liner for maximum protection of blade and user
- Rib back blade design on select blades offers greater cutting control and strength
- Available with plastic shield SafetyLock to securely cover the blade edge before and after use
- Available sterile in carbon and stainless steel and nonsterile in carbon steel
Specifications
| Carbon Steel | |
| Surgical Blade | |
| For Tissue Separation, and Other Procedures that require a Sharp Surgical Blade to Puncture or Cut |