Learn More
Aspen Surgical™ Bard-Parker™ Surgical Blades
Catalog No. 22027982
Change view
Click to view available options
Size:
# 10
# 11
# 15
# 21
# 22
# 25
Sterility:
Non-sterile
Sterile
Handle Type:
Ribbed
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Sterility Handle Type
22027982 # 22 Sterile Ribbed
0891511 # 21 Sterile Ribbed
08914A # 10 Non-sterile Ribbed
08914B # 11 Non-sterile Ribbed
089151B # 22 Non-sterile Ribbed
089151C # 25 Non-sterile Ribbed
089159 # 15 Sterile Ribbed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 22027982 Supplier Aspen Surgical™ Supplier No. 371122
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Offer consistent sharpness, control and strength
  • Each blade packaged in foil pouch with liner for maximum protection of blade and user
  • Rib back blade design on select blades offers greater cutting control and strength
  • Available with plastic shield SafetyLock to securely cover the blade edge before and after use
  • Available sterile in carbon and stainless steel and nonsterile in carbon steel
Specifications
Blade Material Carbon Steel
Product Type Surgical Blade
For Use With (Application) For Tissue Separation, and Other Procedures that require a Sharp Surgical Blade to Puncture or Cut
Handle Type Ribbed
Size # 22
Sterility Sterile