missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAPTA tetraethyl ester, 98+%
CAS: 73630-07-6 | C30H40N2O10 | 588.654 g/mol
$213.60 - $739.66
Chemical Identifiers
| CAS | 73630-07-6 |
|---|---|
| Molecular Formula | C30H40N2O10 |
| Molecular Weight (g/mol) | 588.654 |
| MDL Number | MFCD00532663 |
| InChI Key | OLXCPQDFHUCXBA-UHFFFAOYSA-N |
| Synonym | bapta tetraethyl ester, tetraethyl 2,2',2,2'-ethane-1,2-diylbis oxy bis 2,1-phenylene bis azanetriyl tetraacetate, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid tetraethyl ester, tetraethyl 2,2',2,2'-ethane-1,2-diylbis oxy-2,1-phenylenenitrilo tetraacetate, 2-2-2-2-bis 2-ethoxy-2-oxoethyl amino phenoxy ethoxy-n-2-ethoxy-2-oxoethyl anilino acetic acid ethyl ester, ethyl 2-2-2-2-bis 2-ethoxy-2-oxidanylidene-ethyl amino phenoxy ethoxy phenyl-2-ethoxy-2-oxidanylidene-ethyl amino ethanoate, ethyl 2-2-2-2-bis 2-ethoxy-2-oxoethyl amino phenoxy ethoxy-n-2-ethoxy-2-oxoethyl anilino acetate, ethyl 2-2-2-2-bis 2-ethoxy-2-oxoethyl amino phenoxy ethoxy phenyl 2-ethoxy-2-oxoethyl amino acetate |
| PubChem CID | 339960 |
| IUPAC Name | ethyl 2-[2-[2-[2-[bis(2-ethoxy-2-oxoethyl)amino]phenoxy]ethoxy]-N-(2-ethoxy-2-oxoethyl)anilino]acetate |
| SMILES | CCOC(=O)CN(CC(=O)OCC)C1=CC=CC=C1OCCOC2=CC=CC=C2N(CC(=O)OCC)CC(=O)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2197103
|
Thermo Scientific Chemicals
B2197103 |
1 g |
Each for $213.60
|
|
|||||
|
AAB2197106
|
Thermo Scientific Chemicals
B2197106 |
5 g |
Each for $739.66
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 73630-07-6 | |
| 588.654 | |
| OLXCPQDFHUCXBA-UHFFFAOYSA-N | |
| 339960 | |
| CCOC(=O)CN(CC(=O)OCC)C1=CC=CC=C1OCCOC2=CC=CC=C2N(CC(=O)OCC)CC(=O)OCC |
| C30H40N2O10 | |
| MFCD00532663 | |
| bapta tetraethyl ester, tetraethyl 2,2',2,2'-ethane-1,2-diylbis oxy bis 2,1-phenylene bis azanetriyl tetraacetate, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid tetraethyl ester, tetraethyl 2,2',2,2'-ethane-1,2-diylbis oxy-2,1-phenylenenitrilo tetraacetate, 2-2-2-2-bis 2-ethoxy-2-oxoethyl amino phenoxy ethoxy-n-2-ethoxy-2-oxoethyl anilino acetic acid ethyl ester, ethyl 2-2-2-2-bis 2-ethoxy-2-oxidanylidene-ethyl amino phenoxy ethoxy phenyl-2-ethoxy-2-oxidanylidene-ethyl amino ethanoate, ethyl 2-2-2-2-bis 2-ethoxy-2-oxoethyl amino phenoxy ethoxy-n-2-ethoxy-2-oxoethyl anilino acetate, ethyl 2-2-2-2-bis 2-ethoxy-2-oxoethyl amino phenoxy ethoxy phenyl 2-ethoxy-2-oxoethyl amino acetate | |
| ethyl 2-[2-[2-[2-[bis(2-ethoxy-2-oxoethyl)amino]phenoxy]ethoxy]-N-(2-ethoxy-2-oxoethyl)anilino]acetate |
Specifications
| 73630-07-6 | |
| C30H40N2O10 | |
| 1 g | |
| OLXCPQDFHUCXBA-UHFFFAOYSA-N | |
| ethyl 2-[2-[2-[2-[bis(2-ethoxy-2-oxoethyl)amino]phenoxy]ethoxy]-N-(2-ethoxy-2-oxoethyl)anilino]acetate | |
| 339960 | |
| ≥98% |
| 98°C to 102°C | |
| MFCD00532663 | |
| bapta tetraethyl ester, tetraethyl 2,2',2,2'-ethane-1,2-diylbis oxy bis 2,1-phenylene bis azanetriyl tetraacetate, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid tetraethyl ester, tetraethyl 2,2',2,2'-ethane-1,2-diylbis oxy-2,1-phenylenenitrilo tetraacetate, 2-2-2-2-bis 2-ethoxy-2-oxoethyl amino phenoxy ethoxy-n-2-ethoxy-2-oxoethyl anilino acetic acid ethyl ester, ethyl 2-2-2-2-bis 2-ethoxy-2-oxidanylidene-ethyl amino phenoxy ethoxy phenyl-2-ethoxy-2-oxidanylidene-ethyl amino ethanoate, ethyl 2-2-2-2-bis 2-ethoxy-2-oxoethyl amino phenoxy ethoxy-n-2-ethoxy-2-oxoethyl anilino acetate, ethyl 2-2-2-2-bis 2-ethoxy-2-oxoethyl amino phenoxy ethoxy phenyl 2-ethoxy-2-oxoethyl amino acetate | |
| CCOC(=O)CN(CC(=O)OCC)C1=CC=CC=C1OCCOC2=CC=CC=C2N(CC(=O)OCC)CC(=O)OCC | |
| 588.654 | |
| 588.66 | |
| BAPTA tetraethyl ester |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only