accessibility menu, dialog, popup

UserName

CH3OONOOCH3OONOOH3COOCH3

CAS RN 73630-07-6

IUPAC Name: ethyl 2-{[2-(2-{2-[bis(2-ethoxy-2-oxoethyl)amino]phenoxy}ethoxy)phenyl](2-ethoxy-2-oxoethyl)amino}acetate
Synonyms: ethyl 2-{[2-(2-{2-[bis(2-ethoxy-2-oxoethyl)amino]phenoxy}ethoxy)phenyl](2-ethoxy-2-oxoethyl)amino}acetate
Molecular Weight (g/mol): 588.65
Molecular Formula: C30H40N2O10
InChi Key: OLXCPQDFHUCXBA-UHFFFAOYSA-N
SMILES: CCOC(=O)CN(CC(=O)OCC)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC(=O)OCC)CC(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results