missing translation for 'onlineSavingsMsg'
Learn More

BAPTA tetraethyl ester, 98+%

Catalog No. AAB2197103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2197103 1 g
AAB2197106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2197103 Supplier Thermo Scientific Chemicals Supplier No. B2197103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 73630-07-6 | C30H40N2O10 | 588.654 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 73630-07-6
Molecular Formula C30H40N2O10
Molecular Weight (g/mol) 588.654
MDL Number MFCD00532663
InChI Key OLXCPQDFHUCXBA-UHFFFAOYSA-N
Synonym bapta tetraethyl ester, tetraethyl 2,2',2,2'-ethane-1,2-diylbis oxy bis 2,1-phenylene bis azanetriyl tetraacetate, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid tetraethyl ester, tetraethyl 2,2',2,2'-ethane-1,2-diylbis oxy-2,1-phenylenenitrilo tetraacetate, 2-2-2-2-bis 2-ethoxy-2-oxoethyl amino phenoxy ethoxy-n-2-ethoxy-2-oxoethyl anilino acetic acid ethyl ester, ethyl 2-2-2-2-bis 2-ethoxy-2-oxidanylidene-ethyl amino phenoxy ethoxy phenyl-2-ethoxy-2-oxidanylidene-ethyl amino ethanoate, ethyl 2-2-2-2-bis 2-ethoxy-2-oxoethyl amino phenoxy ethoxy-n-2-ethoxy-2-oxoethyl anilino acetate, ethyl 2-2-2-2-bis 2-ethoxy-2-oxoethyl amino phenoxy ethoxy phenyl 2-ethoxy-2-oxoethyl amino acetate
PubChem CID 339960
IUPAC Name ethyl 2-[2-[2-[2-[bis(2-ethoxy-2-oxoethyl)amino]phenoxy]ethoxy]-N-(2-ethoxy-2-oxoethyl)anilino]acetate
SMILES CCOC(=O)CN(CC(=O)OCC)C1=CC=CC=C1OCCOC2=CC=CC=C2N(CC(=O)OCC)CC(=O)OCC

Specifications

Melting Point 98°C to 102°C
Quantity 1 g
Formula Weight 588.66
Percent Purity ≥98%
Chemical Name or Material BAPTA tetraethyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only