missing translation for 'onlineSavingsMsg'
Learn More

BAPTA, 97%

Catalog No. AAA1319014
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1319014 25 g
AAA1319003 1 g
AAA1319006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1319014 Supplier Thermo Scientific Chemicals Supplier No. A1319014

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 85233-19-8 | C22H24N2O10 | 476.44 g/mol

Acts as a calcium chelatorBAPTA is used as a selective calcium chelating reagent. It is suitable for the spectrophotometric monitoring of calcium levels. It is a lipophilic diesters of the chelating agent used for the inhibition of proteolytic activities of certain metalloproteinases, calpain and transcatheter arterial chemoembolization (TACE).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acts as a calcium chelatorBAPTA is used as a selective calcium chelating reagent. It is suitable for the spectrophotometric monitoring of calcium levels. It is a lipophilic diesters of the chelating agent used for the inhibition of proteolytic activities of certain metalloproteinases, calpain and transcatheter arterial chemoembolization (TACE).

Solubility
Soluble in water and sodium bicarbonate.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 85233-19-8
Molecular Formula C22H24N2O10
Molecular Weight (g/mol) 476.44
MDL Number MFCD00036255
InChI Key FTEDXVNDVHYDQW-UHFFFAOYSA-N
Synonym bapta, bapeta, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid, ca ii-bapta, 1,2-bis-2-aminophenoxy ethane n,n,n',n'-tetraacetic acid, unii-k22ddw77c0, 1,2-bis o-aminophenoxy ethane-n,n,n',n'-tetraacetic acid, bapta tetrasodium salt hydrate, glycine, n,n'-1,2-ethanediylbis oxy-2,1-phenylene bis n-carboxymethyl, 2,2',2,2'-ethane-1,2-diylbis oxy-2,1-phenylenenitrilo tetraacetic acid
PubChem CID 104751
ChEBI CHEBI:60888
IUPAC Name 2-{[2-(2-{2-[bis(carboxymethyl)amino]phenoxy}ethoxy)phenyl](carboxymethyl)amino}acetic acid
SMILES OC(=O)CN(CC(O)=O)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC(O)=O)CC(O)=O

Specifications

Melting Point ∼174°C (decomposition)
Quantity 25 g
Merck Index 14,957
Solubility Information Soluble in water and sodium bicarbonate.
Formula Weight 476.44
Percent Purity 97%
Chemical Name or Material BAPTA
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber MC0423000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only