accessibility menu, dialog, popup

UserName

CAS RN 85233-19-8

IUPAC Name: 2-{[2-(2-{2-[bis(carboxymethyl)amino]phenoxy}ethoxy)phenyl](carboxymethyl)amino}acetic acid
Synonyms: bapta|bapeta
Molecular Weight (g/mol): 476.44
Molecular Formula: C22H24N2O10
InChi Key: FTEDXVNDVHYDQW-UHFFFAOYSA-N
SMILES: OC(=O)CN(CC(O)=O)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC(O)=O)CC(O)=O
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)