Learn More
BAKER H2O-Philic SC-DVB Resin, J.T. Baker™
Catalog No. 02002000
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02002000 100 g
02002002 500 g
02002001 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02002000 Supplier Avantor Supplier No. 811200
STR Cation Exchanger
Specifications
Chemical Name or Material BAKER H2O-Philic SC-DVB Resin
Pore Size 15 μm, 200 Å
Quantity 100 g
Packaging Poly Bottle