missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAIAP2 Mouse anti-Mouse, Human, Clone: CA-A10-5, MilliporeSigma™
Description
BAIAP2 Monoclonal antibody specifically detects BAIAP2 in Mouse, Human samples. It is validated for Dot Blot, Immunocytochemistry, Immunofluorescence, Western Blotting.
Specifications
Specifications
| Antigen | BAIAP2 |
| Applications | Dot Blot, Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | CA-A10-5 |
| Conjugate | Unconjugated |
| Formulation | Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. |
| Host Species | Mouse |
| Immunogen | His-tagged full-length human recombinant Brain-specific angiogenesis inhibitor 1-associated protein 2 (BAIAP2) |
| Purification Method | Protein G purified |
| Quantity | 25 μL |
| Show More |