missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BAG2 Recombinant Rabbit Monoclonal Antibody (23GB7975)

Catalog No. MA552755
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552755 20 μL
MA552754 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552755 Supplier Invitrogen™ Supplier No. MA552755
Only null left

Rabbit Recombinant Monoclonal Antibody

BAG2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF

BAG proteins compete with Hip for binding to the Hsc70/Hsp70 ATPase domain and promote substrate release. All the BAG proteins have an approximately 45-amino acid BAG domain near the C terminus but differ markedly in their N-terminal regions. The predicted BAG2 protein contains 211 amino acids. The BAG domains of BAG1, BAG2, and BAG3 interact specifically with the Hsc70 ATPase domain in vitro and in mammalian cells. All 3 proteins bind with high affinity to the ATPase domain of Hsc70 and inhibit its chaperone activity in a Hip-repressible manner.

Specifications

Antigen BAG2
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB7975
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BAG2
Gene Accession No. O95816, Q91YN9
Gene Alias 2610042A13Rik; BAG cochaperone 2; BAG family molecular chaperone regulator 2; BAG2; BAG-2; BAG-family molecular chaperone regulator-2; BC016230; BCL2 associated athanogene 2; Bcl-2-associated athanogene 2; Bcl2-associated athanogene 2; dJ417I1.2; dJ417I1.2 (BAG-family molecular chaperone regulator 2); KIAA0576; MGC149462; RP3-496N17.2
Gene Symbols BAG2
Host Species Rabbit
Immunogen A synthesized peptide derived from BAG2 (150-211AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213539, 690038, 9532
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less