missing translation for 'onlineSavingsMsg'
Learn More

BAG1 Recombinant Rabbit Monoclonal Antibody (23GB2995)
GREENER_CHOICE

Catalog No. MA553322
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553322 100 μL
MA553323 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553322 Supplier Fisher Chemical Supplier No. MA553322
Only null left

BAG1 Recombinant Rabbit Monoclonal Antibody (23GB2995)

BAG1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Specifications

Antigen BAG1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2995
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Bag1
Gene Accession No. B0K019, Q60739, Q99933
Gene Alias BAG family molecular chaperone regulator 1; Bag1; BAG-1; BCL2 associated athanogene 1; Bcl-2 associating athanogene-1 protein; BCL2-associated athanogene; bcl-2-associated athanogene 1; BCL2-associated athanogene 1; Bcl-2-binding protein; glucocortoid receptor-associated protein RAP46; HAP; Rap46; receptor-associated protein, 46-KD; RP11-326F20.2
Gene Symbols Bag1
Host Species Rabbit
Immunogen A synthesized peptide derived from Bag1 (130-155AA).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12017, 297994, 573
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less