missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAD Mouse anti-Human, Clone:1G5B3, Abnova™
Mouse monoclonal antibody raised against recombinant human BAD.
Supplier: Abnova Corporation MAB16598
Description
The protein encoded by this gene is a member of the BCL-2 family. BCL-2 family members are known to be regulators of programmed cell death. This protein positively regulates cell apoptosis by forming heterodimers with BCL-xL and BCL-2, and reversing their death repressor activity. Proapoptotic activity of this protein is regulated through its phosphorylation. Protein kinases AKT and MAP kinase, as well as protein phosphatase calcineurin were found to be involved in the regulation of this protein. Alternative splicing of this gene results in two transcript variants which encode the same isoform. [provided by RefSeq]Specifications
| BAD | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
| BAD | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 1G5B3 | |
| BCL2-associated agonist of cell death | |
| In PBS (0.05% sodium azide) | |
| BBC2, BCL2L8 | |
| Recombinant protein corresponding to amino acid 1-168 of human BAD from E. coli. | |
| 572 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |