missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Yeast Extract, technical
GREENER_CHOICE

Suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications.

Supplier:  Gibco™ 288610

 View more versions of this product

Catalog No. DF0886081


Only null left

Description

Description

  • Concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells derived from primary grown baker's yeast
  • These animal origin-free products are suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications
  • Yeast extract provides essential water-soluble vitamins, amino acids, peptides, and carbohydrates to any medium formulation
  • Bacto Yeast Extract is an important ingredient for the microbiological assay of vitamins
  • B factor, a growth substance necessary for the production of rifampin in a Nocardia species, can be isolated from yeast extract
  • Developed to provide products priced for the biotechnology market with acceptable clarity and growth-promoting characteristics
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
Specifications

Chemical Identifiers

Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Bacterial Media
Bacto
Mammalian, Insect, Microbial
Dehydrated culture media
Animal origin-free

Specifications

LUO, MUO
Mammalian, Insect, Microbial
Bacterial Media
Dehydrated culture media
10 kg
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Powder
Pail
Bacto
Animal origin-free
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 1825 Days

SDS
Documents

Documents

Promotions

Promotions