missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ TC Yeastolate
GREENER_CHOICE

Catalog No. DF5577155
Change view
Click to view available options
Quantity:
100 g
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF5577155 100 g
B55771 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DF5577155 Supplier Gibco™ Supplier No. 255772

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bacto TC Yeastolate is an animal origin-free hydrolysate from the water-soluble portion of autolyzed yeast. This supplement contains a mixture of peptides, amino acids, carbohydrates (simple and complex), nucleosides, and vitamins. Derived from the same yeast strain as Difco TC Yeastolate, ultra-filtered (UF), this product has a slightly different nutritional profile because it was not ultra-filtered.

Bacto TC Yeastolate is an animal origin-free hydrolysate from the water-soluble portion of autolyzed yeast. This supplement contains a mixture of peptides, amino acids, carbohydrates (simple and complex), nucleosides, and vitamins. Derived from the same yeast strain as Difco TC Yeastolate, ultra-filtered (UF), this product has a slightly different nutritional profile because it was not ultra-filtered. Bacto TC Yeastolate should be considered a suitable alternative to Difco TC Yeastolate, ultra-filtered (UF), for supplementing a variety of CHO-based applications.

Customers have used this supplement for CHO-based production of biotherapeutic monoclonal antibodies used in oncological and rheumatological therapeutics and recombinant proteins. Additionally, customers have used this supplement in combination with a soy supplement, such as Difco Phytone, ultra-filtered (UF), to provide an optimal titer performance in mammalian cell based processes by providing additional vitamins and carbohydrates.

Specifications

Content And Storage Store at 2°C to 25°C in cool, dry conditions in well-sealed containers.
Cell Type Bacteria
Product Type TC Yeastolate
Product Line Bacto
Quantity 100 g
Source Animal origin-free