missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ TC Lactalbumin Hydrolysate
GREENER_CHOICE

Catalog No. DF5996170
Change view
Click to view available options
Quantity:
500 g
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF5996170 500 g
259961 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DF5996170 Supplier Gibco™ Supplier No. 259962

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An animal origin peptone from the enzymatically hydrolyzed protein portion of milk whey.

This supplement is a mixture of peptides, amino acids, and carbohydrates, both simple and complex. Customers have successfully used Gibco™ Bacto™ TC Lactalbumin Hydrolysate to support the growth of many mammalian, bacterial, and insect cell types, and it has been used to produce both recombinant proteins and animal health vaccines. Customers have also used this supplement as an amino acid supplement for mammalian cell culture media.

Specifications

Content And Storage Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.
Culture Type Mammalian, Insect, Microbial
Form Powder
Product Type Dehydrated Culture Media
For Use With (Application) Mammalian, bacterial, insect cell types, recombinant proteins and animal health vaccines
Product Line Bacto
Quantity 500 g
Source Animal Origin