missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Soytone
GREENER_CHOICE

Catalog No. DF0436175
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0436175 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0436175 Supplier Gibco™ Supplier No. 243620

An enzymatic digest of soy that uses an animal origin (AO) (porcine) enzyme for digestion.

  • An enzymatic digest of soy that uses an animal origin (AO) (porcine) enzyme for digestion
  • The naturally high concentrations of vitamins and carbohydrates found in soy make Bacto Soytone a nutritious carbohydrate source for those not concerned about the processing step involving an animal-based material
  • This supplement offers a unique nutritional profile as compared to other soy products
  • It can be a valuable alternative to consider for supplementing both mammalian and microbial processes
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C

Specifications

Certifications/Compliance LUO
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Dehydrated Culture Media
Format Bottle
For Use With (Application) Enzymatic Hydrolysate of Soybean meal
Product Line Bacto
Quantity 500 g
Source Animal origin, porcine